Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myh11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024448.1 | NP_001019619.1 | 8 - 9 | 998 | 70 | |
XM_021476875.1 | XP_021332550.1 | 9 - 10 | 70 | |||
XM_021476876.1 | XP_021332551.1 | 9 - 10 | 70 | |||
GenBank mRNA | BC162152.1 | - | 8 - 9 | 70 | ||
DQ022376.1 | - | 8 - 9 | 998 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |