Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025551.1 | NP_001020722.1 | 3 - 4 | 1417 | 72 | |
XM_005160586.3 | XP_005160643.1 | 4 - 5 | 1986 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |