Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itgb1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034987.1 | NP_001030159.1 | 362 | 112 | ||
GenBank mRNA | BC162706.1 | - | 459 | 112 | ||
DQ149102.1 | - | 362 | 112 | |||
HQ538840.1 | - | 632 | 112 |
Gene Symbol: LOC110437890 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021466326.1 | XP_021322001.1 | 4 - 5 | 702 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |