Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uba52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037113.1 | NP_001032190.1 | 4 - 5 | 344 | 83 | |
GenBank mRNA | BC076111.1 | - | 1 - 2 | 174 | 83 | |
BC105746.1 | - | 4 - 5 | 352 | 83 | ||
BC107517.1 | - | 4 - 5 | 344 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |