Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037686.1 | NP_001032775.1 | 7 - 8 | 1672 | 96 | |
GenBank mRNA | AY928519.1 | - | 7 - 8 | 1672 | 96 | |
BC163582.1 | - | 7 - 8 | 1747 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |