Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abcg2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042772.1 | NP_001036237.1 | 11 - 12 | 1609 | 104 | |
GenBank mRNA | DQ223861.1 | - | 11 - 12 | 1609 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |