Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn4bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077573.1 | NP_001071041.1 | 4 - 5 | 1034 | 80 | |
GenBank mRNA | BC146712.1 | - | 4 - 5 | 1033 | 80 | |
BC162278.1 | - | 4 - 5 | 914 | 80 | ||
BC162294.1 | - | 4 - 5 | 914 | 80 | ||
DQ471306.1 | - | 3 - 4 | 488 | 80 | ||
DQ489732.1 | - | 4 - 5 | 1034 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |