Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077737.1 | NP_001071205.1 | 13 - 14 | 1752 | 100 | |
XM_005161204.4 | XP_005161261.1 | 16 - 17 | 2041 | 100 | ||
XM_009295332.3 | XP_009293607.1 | 16 - 17 | 1961 | 100 | ||
GenBank mRNA | BC125837.1 | - | 13 - 14 | 1752 | 100 | |
BC165661.1 | - | 13 - 14 | 1539 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |