Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: creb3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080651.2 | NP_001074120.1 | 8 - 9 | 1353 | 137 | |
XM_021475142.1 | XP_021330817.1 | 7 - 8 | 1043 | 137 | ||
GenBank mRNA | BC129318.1 | - | 8 - 9 | 1355 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |