Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acot13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080803.1 | NP_001074272.1 | 2 - 3 | 282 | 72 | |
XM_005159635.4 | XP_005159692.1 | 2 - 3 | 375 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Danio rerio Probe spans exons |