Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taar14h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083093.1 | NP_001076562.1 | 1 | 549 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |