Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hspa13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089479.1 | NP_001082948.1 | 2 - 3 | 422 | 148 | |
GenBank mRNA | BC139562.1 | - | 2 - 3 | 422 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |