Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099237.2 | NP_001092707.2 | 3 - 4 | 418 | 90 | |
XM_005161956.4 | XP_005162013.1 | 3 - 4 | 412 | 90 | ||
XM_005161957.4 | XP_005162014.1 | 3 - 4 | 412 | 90 | ||
GenBank mRNA | BC070016.1 | - | 2 - 3 | 361 | 90 | |
BC129148.1 | - | 2 - 3 | 361 | 90 | ||
BC142793.1 | - | 3 - 4 | 418 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |