Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl6ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100074.1 | NP_001093544.1 | 4 - 5 | 1126 | 103 | |
XM_005171137.3 | XP_005171194.1 | 4 - 5 | 1145 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Danio rerio Probe spans exons |