Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qdpra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110469.1 | NP_001103939.1 | 4 - 5 | 452 | 134 | |
GenBank mRNA | BC115316.1 | - | 4 - 5 | 452 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |