Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drosha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110472.1 | NP_001103942.1 | 21 - 22 | 2978 | 134 | |
GenBank mRNA | BC133134.1 | - | 21 - 22 | 2978 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |