Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drosha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110472.1 | NP_001103942.1 | 25 - 26 | 3358 | 71 | |
GenBank mRNA | BC133134.1 | - | 25 - 26 | 3358 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |