Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: skia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130935.3 | NP_571010.3 | 5 - 6 | 2140 | 69 | |
GenBank mRNA | AF060117.1 | - | 5 - 6 | 2145 | 69 | |
BC117641.1 | - | 5 - 6 | 2140 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |