Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gli2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130967.1 | NP_571042.1 | 6 - 7 | 1168 | 90 | |
GenBank mRNA | AF085746.2 | - | 6 - 7 | 1168 | 90 | |
AF348156.1 | - | 3 - 4 | 559 | 90 | ||
BC163542.1 | - | 6 - 7 | 1159 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |