Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: noto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131055.1 | NP_571130.1 | 1 - 2 | 366 | 93 | |
GenBank mRNA | BC162344.1 | - | 1 - 2 | 366 | 93 | |
L48017.1 | - | 1 - 2 | 366 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |