Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otx2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131251.1 | NP_571326.1 | 3 - 4 | 509 | 76 | |
XM_005158883.4 | XP_005158940.1 | 4 - 5 | 548 | 76 | ||
XM_009293247.3 | XP_009291522.1 | 3 - 4 | 472 | 76 | ||
GenBank mRNA | BC115165.1 | - | 2 - 3 | 506 | 76 | |
BC164653.1 | - | 2 - 3 | 280 | 76 | ||
D26173.1 | - | 2 - 3 | 324 | 76 | ||
U14592.1 | - | 3 - 4 | 509 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |