Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131357.1 | NP_571432.1 | 9 - 10 | 1125 | 105 | |
GenBank mRNA | AF132084.1 | - | 9 - 10 | 1125 | 105 | |
BC162211.1 | - | 9 - 10 | 1067 | 105 | ||
BC162544.1 | - | 9 - 10 | 1067 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Danio rerio Probe spans exons |