Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131713.2 | NP_571788.2 | 3 - 4 | 523 | 128 | |
GenBank mRNA | AF247823.1 | - | 1 - 2 | 103 | 128 | |
BC095063.1 | - | 3 - 4 | 456 | 128 | ||
BC164464.1 | - | 2 - 3 | 387 | 128 | ||
CU468784.1 | - | 2 - 3 | 353 | 128 | ||
EE689153.1 | - | 3 - 4 | 580 | 128 | ||
EE697401.1 | - | 2 - 3 | 206 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |