Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198121.2 | NP_937754.2 | 2 - 3 | 554 | 139 | |
GenBank mRNA | AF251282.1 | - | 2 - 3 | 308 | 139 | |
AY391410.1 | - | 2 - 3 | 426 | 139 | ||
AY930110.1 | - | 2 - 3 | 308 | 139 | ||
AY930111.1 | - | 2 - 3 | 308 | 139 | ||
BC053242.1 | - | 2 - 3 | 501 | 139 | ||
BG737536.1 | - | 2 - 3 | 509 | 139 | ||
EH551253.1 | - | 2 - 3 | 593 | 139 | ||
EH570664.1 | - | 2 - 3 | 678 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |