Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prrx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200050.1 | NP_956344.1 | 3 - 4 | 964 | 75 | |
GenBank mRNA | BC064671.1 | - | 3 - 4 | 964 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |