Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC046008.1 | - | 5 - 6 | 609 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |