Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agfg1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200588.2 | NP_956882.2 | 4 - 5 | 592 | 71 | |
XM_005163316.4 | XP_005163373.1 | 4 - 5 | 687 | 71 | ||
GenBank mRNA | BC056733.1 | - | 4 - 5 | 592 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |