Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agfg1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200588.2 | NP_956882.2 | 6 - 7 | 858 | 59 | |
GenBank mRNA | BC056733.1 | - | 6 - 7 | 858 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |