Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adam8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200637.1 | NP_956931.1 | 15 - 16 | 1651 | 151 | |
GenBank mRNA | BC057428.1 | - | 15 - 16 | 1651 | 151 | |
BC165674.1 | - | 15 - 16 | 1649 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |