Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adam8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200637.1 | NP_956931.1 | 7 - 8 | 667 | 94 | |
GenBank mRNA | BC057428.1 | - | 7 - 8 | 667 | 94 | |
BC165674.1 | - | 7 - 8 | 665 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |