Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap2m1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201026.2 | NP_957320.1 | 8 - 9 | 1142 | 65 | |
XM_005159256.4 | XP_005159313.1 | 9 - 10 | 1193 | 65 | ||
XM_009293860.3 | XP_009292135.1 | 10 - 11 | 1208 | 65 | ||
GenBank mRNA | BC049515.1 | - | 8 - 9 | 1144 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |