Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkcba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201029.1 | NP_957323.1 | 16 - 17 | 1967 | 87 | |
GenBank mRNA | BC054603.1 | - | 16 - 17 | 1967 | 87 | |
BC171680.1 | - | 16 - 17 | 1931 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |