Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdh10b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201331.1 | NP_958488.1 | 2 - 3 | 627 | 83 | |
GenBank mRNA | AM422096.1 | - | 1 - 2 | 196 | 83 | |
BC054596.1 | - | 2 - 3 | 627 | 83 | ||
BC164370.1 | - | 2 - 3 | 537 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |