Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ldb3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201505.1 | NP_958913.1 | 7 - 8 | 713 | 71 | |
GenBank mRNA | BC045908.1 | - | 7 - 8 | 713 | 71 | |
BC164050.1 | - | 6 - 7 | 611 | 71 | ||
DQ012165.1 | - | 6 - 7 | 577 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |