Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jade3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201652.2 | NP_970614.2 | 3 - 4 | 268 | 73 | |
GenBank mRNA | BC045468.1 | - | 3 - 4 | 268 | 73 | |
BC165248.1 | - | 1 - 2 | 89 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |