Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bnip3lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205571.2 | NP_991134.2 | 2 - 3 | 303 | 119 | |
GenBank mRNA | AF493987.1 | - | 2 - 3 | 273 | 119 | |
BC091797.1 | - | 2 - 3 | 303 | 119 | ||
BC165026.1 | - | 2 - 3 | 305 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |