Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gria1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205730.2 | NP_991293.1 | 15 - 16 | 2902 | 82 | |
XM_005161339.4 | XP_005161396.1 | 15 - 16 | 2902 | 82 | ||
GenBank mRNA | AF525742.1 | - | 15 - 16 | 2687 | 82 | |
BC162147.1 | - | 15 - 16 | 2623 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |