Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arrb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214681.1 | NP_999846.1 | 8 - 9 | 716 | 103 | |
XM_009305269.3 | XP_009303544.1 | 8 - 9 | 754 | 103 | ||
GenBank mRNA | BC057519.1 | - | 8 - 9 | 716 | 103 | |
BC165790.1 | - | 8 - 9 | 656 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |