Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp13a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001403.2 | NP_001001403.2 | 17 - 18 | 2560 | 78 | |
GenBank mRNA | BC055391.1 | - | 17 - 18 | 2560 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |