Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sgcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001816.4 | NP_001001816.2 | 7 - 8 | 995 | 66 | |
XM_009295594.3 | XP_009293869.1 | 7 - 8 | 1154 | 66 | ||
GenBank mRNA | AY398418.1 | - | 6 - 7 | 705 | 66 | |
BC095349.1 | - | 7 - 8 | 66 | |||
BC164654.1 | - | 6 - 7 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |