Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fntb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002128.1 | NP_001002128.1 | 1 - 2 | 163 | 99 | |
XM_021468366.1 | XP_021324041.1 | 1 - 2 | 187 | 99 | ||
GenBank mRNA | BC071443.1 | - | 1 - 2 | 163 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |