Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fntb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002128.1 | NP_001002128.1 | 9 - 10 | 983 | 69 | |
XM_021468366.1 | XP_021324041.1 | 8 - 9 | 920 | 69 | ||
GenBank mRNA | BC071443.1 | - | 9 - 10 | 983 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |