Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab3db | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002679.1 | NP_001002679.1 | 2 - 3 | 442 | 125 | |
XM_005168476.4 | XP_005168533.1 | 2 - 3 | 344 | 125 | ||
GenBank mRNA | BC075784.1 | - | 2 - 3 | 442 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |