Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005601.2 | NP_001005601.1 | 4 - 5 | 427 | 59 | |
GenBank mRNA | BC082297.1 | - | 4 - 5 | 431 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |