Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tap2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006594.1 | NP_001006594.1 | 5 - 6 | 1152 | 70 | |
GenBank mRNA | BC163447.1 | - | 5 - 6 | 1186 | 70 | |
BC163453.1 | - | 5 - 6 | 1186 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |