Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007415.1 | NP_001007416.1 | 12 - 13 | 1395 | 129 | |
XM_005171163.4 | XP_005171220.1 | 12 - 13 | 1256 | 129 | ||
XM_021480037.1 | XP_021335712.1 | 12 - 13 | 1374 | 129 | ||
GenBank mRNA | BC085370.1 | - | 12 - 13 | 1395 | 129 | |
BC165850.1 | - | 11 - 12 | 1174 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |