Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007415.1 | NP_001007416.1 | 18 - 19 | 1936 | 67 | |
XM_005171163.4 | XP_005171220.1 | 18 - 19 | 1797 | 67 | ||
XM_021480037.1 | XP_021335712.1 | 18 - 19 | 1915 | 67 | ||
GenBank mRNA | BC085370.1 | - | 18 - 19 | 1936 | 67 | |
BC165850.1 | - | 17 - 18 | 1715 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |