Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cx23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013546.1 | NP_001013564.1 | 2 - 3 | 342 | 68 | |
XM_005160685.3 | XP_005160742.1 | 2 - 3 | 413 | 68 | ||
XM_021468416.1 | XP_021324091.1 | 3 - 4 | 336 | 68 | ||
GenBank mRNA | BC091468.1 | - | 2 - 3 | 342 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |