Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kif7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014816.1 | NP_001014816.1 | 17 - 18 | 3716 | 127 | |
XM_005166379.4 | XP_005166436.1 | 18 - 19 | 3887 | 127 | ||
GenBank mRNA | AY954727.1 | - | 17 - 18 | 3716 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons |