Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ism2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020509.1 | NP_001018345.1 | 3 - 4 | 588 | 63 | |
GenBank mRNA | AY856372.1 | - | 3 - 4 | 588 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |